Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rbc-1
Entrez Gene ID
181558
Gene Name
protein-coding
Gene Aliases
CELE_F54E4.1
Chromosome Location
Chr.X: 14611374 - 14624894 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270317.4 NP_001257246.2 20 - 21 5366 86
NM_001373583.1 NP_001360775.1 20 - 21 5333 86
NM_001381151.1 NP_001368758.1 20 - 21 5349 86
NM_001381152.1 NP_001368759.1 20 - 21 5349 86
NM_001381153.1 NP_001368756.1 20 - 21 5316 86
NM_001381154.1 NP_001368757.1 20 - 21 5316 86
NM_001381155.1 NP_001368146.1 20 - 21 5343 86
NM_001381156.2 NP_001368760.1 20 - 21 5339 86

View Full Product Details