Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
syg-2
Entrez Gene ID
181561
Gene Name
protein-coding
Gene Aliases
CELE_C26G2.1
Chromosome Location
Chr.X: 14654968 - 14662865 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322663.4 NP_001309674.1 17 - 18 2706 119
NM_001322664.3 NP_001309467.1 16 - 17 2667 119
NM_001322665.4 NP_001309678.1 12 - 13 1778 119
NM_001322667.4 NP_001309677.1 6 - 7 1241 119
NM_001322668.4 NP_001309676.1 6 - 7 1154 119
NM_001383675.1 16 - 17 2660 119

View Full Product Details