Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
syg-2
Entrez Gene ID
181561
Gene Name
protein-coding
Gene Aliases
CELE_C26G2.1
Chromosome Location
Chr.X: 14654968 - 14662865 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322663.4 NP_001309674.1 5 - 6 882 76
NM_001322664.3 NP_001309467.1 5 - 6 882 76

View Full Product Details