Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cutl-3
Entrez Gene ID
184721
Gene Name
ZP domain-containing protein
Gene Aliases
CELE_F20D1.8
Chromosome Location
Chr.X: 14994752 - 14999202 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078091.5 NP_510492.2 6 - 7 744 80

View Full Product Details