Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxa-96
Entrez Gene ID
186676
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_H03G16.4
Chromosome Location
Chr.X: 15058369 - 15060403 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078101.4 NP_510502.1 3 - 4 309 91

View Full Product Details