Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srg-64
Entrez Gene ID
191395
Gene Name
Serpentine receptor class gamma
Gene Aliases
CELE_ZK678.6
Chromosome Location
Chr.X: 15736844 - 15740626 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078188.2 NP_510589.2 3 - 4 656 77

View Full Product Details