Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
osm-1
Entrez Gene ID
181715
Gene Name
Intraflagellar transport protein osm-1
Gene Aliases
CELE_T27B1.1
Chromosome Location
Chr.X: 16540674 - 16551211 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078280.8 NP_510681.4 3 - 4 1039 89
GenBank mRNA DQ360811.1 - 3 - 4 323 89

View Full Product Details