Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nhr-1
Entrez Gene ID
181783
Gene Name
protein-coding
Gene Aliases
CELE_R09G11.2
Chromosome Location
Chr.X: 17514063 - 17528197 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029684.6 NP_001024855.1 5 - 6 515 70
NM_001381184.1 NP_001367802.1 4 - 5 423 70
GenBank mRNA AY204163.1 - 5 - 6 499 70
AY204164.1 - 5 - 6 513 70

View Full Product Details