Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tpxl-1
Entrez Gene ID
171786
Gene Name
protein-coding
Gene Aliases
CELE_Y39G10AR.12
Chromosome Location
Chr.I: 2297280 - 2304397 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170821.6 NP_740805.1 3 - 4 1273 68
NM_170822.8 NP_740806.1 3 - 4 1277 68
GenBank mRNA DQ178245.1 - 3 - 4 1273 68
DQ178246.1 - 3 - 4 1273 68

View Full Product Details