Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-64
Entrez Gene ID
260292
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_Y39G10AR.22
Chromosome Location
Chr.I: 2327989 - 2330162 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170823.2 NP_740807.1 2 - 3 638 63

View Full Product Details