Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nsf-1
Entrez Gene ID
266842
Gene Name
protein-coding
Gene Aliases
CELE_H15N14.2
Chromosome Location
Chr.I: 7766532 - 7772231 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083134.4 NP_001076603.1 2 - 3 142 57

View Full Product Details