Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tbc-6
Entrez Gene ID
182185
Gene Name
Growth hormone-regulated TBC protein 6
Gene Aliases
CELE_C04A2.1
Chromosome Location
Chr.II: 6848060 - 6851534 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026738.6 NP_001021909.1 2 - 3 222 95

View Full Product Details