Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unc-53
Entrez Gene ID
174693
Gene Name
protein-coding
Gene Aliases
CELE_F45E10.1
Chromosome Location
Chr.II: 11058047 - 11089456 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026998.5 NP_001022169.1 4 - 5 277 95
NM_001026999.8 NP_001022170.1 4 - 5 277 95
NM_001027000.6 NP_001022171.1 1 - 2 72 95
NM_001267342.3 NP_001254271.1 4 - 5 277 95
NM_001267343.3 NP_001254272.1 4 - 5 277 95
NM_171039.3 NP_741045.1 4 - 5 318 95

View Full Product Details