Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
frm-8
Entrez Gene ID
175428
Gene Name
protein-coding
Gene Aliases
CELE_H09G03.2
Chromosome Location
Chr.III: 3200795 - 3219150 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171096.6 NP_741109.1 11 - 12 2472 73
NM_171097.5 NP_741110.1 10 - 11 2360 73

View Full Product Details