Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
frm-8
Entrez Gene ID
175428
Gene Name
protein-coding
Gene Aliases
CELE_H09G03.2
Chromosome Location
Chr.III: 3200795 - 3219150 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065271.6 NP_497672.2 2 - 3 224 74
NM_171096.6 NP_741109.1 3 - 4 348 74
NM_171097.5 NP_741110.1 2 - 3 236 74

View Full Product Details