Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sax-2
Entrez Gene ID
175697
Gene Name
protein-coding
Gene Aliases
CELE_F21H11.2
Chromosome Location
Chr.III: 5109701 - 5130676 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171117.5 NP_741131.2 22 - 23 5817 144

View Full Product Details