Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pqe-1
Entrez Gene ID
175731
Gene Name
protein-coding
Gene Aliases
CELE_F52C9.8
Chromosome Location
Chr.III: 5311240 - 5326509 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047389.4 NP_001040854.1 8 - 9 986 92
NM_001382869.1 10 - 11 4122 92
NM_065734.8 NP_498135.2 11 - 12 3918 92
NM_065735.6 NP_498136.2 8 - 9 981 92

View Full Product Details