Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-39
Entrez Gene ID
176068
Gene Name
Homeobox protein lin-39
Gene Aliases
CELE_C07H6.7
Chromosome Location
Chr.III: 7528641 - 7536649 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025993.4 NP_001021164.1 5 - 6 733 133
GenBank mRNA L19248.1 - 4 - 5 778 133
L19639.1 - 5 - 6 716 133

View Full Product Details