Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unc-16
Entrez Gene ID
176349
Gene Name
protein-coding
Gene Aliases
CELE_ZK1098.10
Chromosome Location
Chr.III: 9546258 - 9557450 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027810.5 NP_001022981.1 13 - 14 2391 61
NM_001129275.4 NP_001122747.1 3 - 4 494 61
NM_001268079.4 NP_001255008.1 14 - 15 2490 61
NM_001268080.3 NP_001255009.1 9 - 10 1580 61
NM_001306679.3 NP_001293608.1 7 - 8 1376 61
NM_001306687.3 NP_001293616.1 8 - 9 1394 61
NM_171221.7 NP_741263.3 15 - 16 2482 61
GenBank mRNA AF424978.1 - 13 - 14 2327 61

View Full Product Details