Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dpf-7
Entrez Gene ID
177250
Gene Name
protein-coding
Gene Aliases
CELE_R11E3.7
Chromosome Location
Chr.IV: 4799384 - 4802594 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380209.2 NP_001367922.1 6 - 7 1574 70
NM_171328.7 NP_741386.2 5 - 6 1618 70

View Full Product Details