Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lag-1
Entrez Gene ID
177373
Gene Name
protein-coding
Gene Aliases
CELE_K08B4.1
Chromosome Location
Chr.IV: 6095407 - 6110655 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171350.7 NP_741410.1 2 - 3 140 67
GenBank mRNA U49794.1 - 2 - 3 138 67

View Full Product Details