Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lag-1
Entrez Gene ID
177373
Gene Name
protein-coding
Gene Aliases
CELE_K08B4.1
Chromosome Location
Chr.IV: 6095407 - 6110655 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028107.5 NP_001023278.1 3 - 4 203 111
NM_001306811.4 NP_001293740.1 1 - 2 69 111
NM_001380259.1 NP_001368385.1 1 - 2 62 111
NM_171350.7 NP_741410.1 3 - 4 209 111
GenBank mRNA U49794.1 - 3 - 4 207 111

View Full Product Details