Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ari-1.1
Entrez Gene ID
172284
Gene Name
RBR-type E3 ubiquitin transferase
Gene Aliases
CELE_C27A12.8
Chromosome Location
Chr.I: 6058546 - 6060953 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059348.6 NP_491749.2 5 - 6 1117 139

View Full Product Details