Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rps-17
Entrez Gene ID
172313
Gene Name
40S ribosomal protein S17
Gene Aliases
CELE_T08B2.10
Chromosome Location
Chr.I: 6219884 - 6220766 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059394.8 NP_491795.1 1 - 2 172 68

View Full Product Details