Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-37
Entrez Gene ID
183583
Gene Name
TWiK family of potassium channels
Gene Aliases
CELE_C48E7.9
Chromosome Location
Chr.I: 6269420 - 6271541 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059409.4 NP_491810.2 3 - 4 297 109

View Full Product Details