Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bli-4
Entrez Gene ID
172333
Gene Name
protein-coding
Gene Aliases
CELE_K04F10.4
Chromosome Location
Chr.I: 6341311 - 6356680 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026369.6 NP_001021540.1 4 - 5 674 107
NM_001026372.7 NP_001021543.1 4 - 5 680 107
NM_001316849.3 NP_001303778.1 4 - 5 680 107
NM_001316850.3 NP_001303779.1 4 - 5 676 107
NM_001316852.3 NP_001303781.1 4 - 5 676 107
NM_001372959.2 NP_001360008.1 5 - 5 681 107
NM_001380517.2 NP_001367786.1 4 - 5 662 107
NM_001380518.2 NP_001367785.1 5 - 6 724 107
NM_001380521.1 NP_001368583.1 3 - 4 516 107
NM_001380523.1 NP_001367787.1 3 - 4 516 107

View Full Product Details