Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rbx-2
Entrez Gene ID
172344
Gene Name
RING-type domain-containing protein
Gene Aliases
CELE_R10A10.2
Chromosome Location
Chr.I: 6420659 - 6421936 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059448.6 NP_491849.1 1 - 2 135 63

View Full Product Details