Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
knl-2
Entrez Gene ID
172350
Gene Name
Kinetochore null protein 2
Gene Aliases
CELE_K06A5.4
Chromosome Location
Chr.I: 6460878 - 6464286 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380547.1 NP_001367697.1 3 - 4 851 82

View Full Product Details