Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dbr-1
Entrez Gene ID
172357
Gene Name
Lariat debranching enzyme
Gene Aliases
CELE_C55B7.8
Chromosome Location
Chr.I: 6490716 - 6492727 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059467.6 NP_491868.2 4 - 5 937 67

View Full Product Details