Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pqn-20
Entrez Gene ID
172402
Gene Name
Prion-like-(Q/N-rich)-domain-bearing protein
Gene Aliases
CELE_C37A2.2
Chromosome Location
Chr.I: 6790978 - 6798161 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059544.7 NP_491945.2 3 - 4 463 129

View Full Product Details