Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpb-5
Entrez Gene ID
172412
Gene Name
DNA-directed RNA polymerases I, II, and III subunit RPABC1
Gene Aliases
CELE_H27M09.2
Chromosome Location
Chr.I: 6850409 - 6851356 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059560.4 NP_491961.1 1 - 2 77 128

View Full Product Details