Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pfd-6
Entrez Gene ID
172474
Gene Name
Probable prefoldin subunit 6
Gene Aliases
CELE_F21C3.5
Chromosome Location
Chr.I: 7282964 - 7283818 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059657.5 NP_492058.1 2 - 3 207 80

View Full Product Details