Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ztf-2
Entrez Gene ID
184429
Gene Name
C2H2-type domain-containing protein
Gene Aliases
CELE_F13G3.1
Chromosome Location
Chr.I: 7291272 - 7295774 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383327.1 2 - 3 739 109

View Full Product Details