Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glb-8
Entrez Gene ID
182935
Gene Name
GLOBIN domain-containing protein
Gene Aliases
CELE_C26C6.7
Chromosome Location
Chr.I: 7509569 - 7512852 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136283.3 NP_001129755.1 5 - 6 807 87

View Full Product Details