Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rrf-1
Entrez Gene ID
172523
Gene Name
protein-coding
Gene Aliases
CELE_F26A3.8
Chromosome Location
Chr.I: 7643426 - 7649777 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001263626.3 NP_001250555.1 10 - 11 2399 121
NM_001263627.3 NP_001250556.1 9 - 10 2146 121
GenBank mRNA AF159144.1 - 10 - 11 2399 121

View Full Product Details