Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
apb-3
Entrez Gene ID
172553
Gene Name
protein-coding
Gene Aliases
CELE_R11A5.1
Chromosome Location
Chr.I: 7853750 - 7857855 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059769.5 NP_492170.2 1 - 2 75 148
NM_059770.5 NP_492171.2 1 - 2 75 148

View Full Product Details