Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nmy-2
Entrez Gene ID
172562
Gene Name
Non-muscle MYosin
Gene Aliases
CELE_F20G4.3
Chromosome Location
Chr.I: 7926599 - 7934760 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059785.9 NP_492186.3 9 - 10 3484 59

View Full Product Details