Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
jph-1
Entrez Gene ID
266843
Gene Name
JunctoPHilin
Gene Aliases
CELE_T22C1.7
Chromosome Location
Chr.I: 7946179 - 7951593 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059792.8 NP_492193.2 3 - 4 603 66

View Full Product Details