Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
abts-1
Entrez Gene ID
172613
Gene Name
protein-coding
Gene Aliases
CELE_F52B5.1
Chromosome Location
Chr.I: 8296917 - 8307572 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136304.4 NP_001129776.1 2 - 3 601 87
NM_001380825.1 NP_001366789.1 2 - 3 124 87
NM_001380826.1 NP_001367194.1 1 - 2 200 87
GenBank mRNA AY887903.1 - 2 - 3 146 87
DQ178237.1 - 1 - 2 282 87

View Full Product Details