Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aph-1
Entrez Gene ID
172747
Gene Name
Gamma-secretase subunit aph-1
Gene Aliases
CELE_VF36H2L.1
Chromosome Location
Chr.I: 9232066 - 9233560 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060068.6 NP_492469.1 2 - 3 279 147

View Full Product Details