Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rba-1
Entrez Gene ID
172801
Gene Name
Probable histone-binding protein rba-1
Gene Aliases
CELE_K07A1.11
Chromosome Location
Chr.I: 9616508 - 9618107 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060150.5 NP_492551.1 3 - 4 741 89

View Full Product Details