Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-53
Entrez Gene ID
172802
Gene Name
Probable histone-binding protein lin-53
Gene Aliases
CELE_K07A1.12
Chromosome Location
Chr.I: 9618196 - 9619760 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060151.9 NP_492552.1 2 - 3 748 72
GenBank mRNA AF116530.1 - 2 - 3 770 72

View Full Product Details