The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128952.3 | NP_001122424.1 | 5 - 6 | 797 | 117 |
NM_060282.6 | NP_492683.1 | 5 - 6 | 797 | 117 |