Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
acs-13
Entrez Gene ID
171643
Gene Name
protein-coding
Gene Aliases
CELE_Y65B4BL.5
Chromosome Location
Chr.I: 511044 - 519646 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058343.5 NP_490744.1 1 - 2 42 60

View Full Product Details