Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
acs-13
Entrez Gene ID
171643
Gene Name
protein-coding
Gene Aliases
CELE_Y65B4BL.5
Chromosome Location
Chr.I: 511044 - 519646 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306267.3 NP_001293196.1 5 - 6 1903 91
NM_001379780.1 NP_001368382.1 4 - 5 1753 91
NM_058343.5 NP_490744.1 5 - 6 1885 91

View Full Product Details