Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cpz-1
Entrez Gene ID
171829
Gene Name
protein-coding
Gene Aliases
CELE_F32B5.8
Chromosome Location
Chr.I: 2686640 - 2689493 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377810.2 NP_001364831.1 1 - 2 95 74
NM_058622.6 NP_491023.2 4 - 5 581 74

View Full Product Details