Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
itx-1
Entrez Gene ID
171846
Gene Name
protein-coding
Gene Aliases
CELE_W03D8.6
Chromosome Location
Chr.I: 2802803 - 2815830 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001262315.4 NP_001249244.2 12 - 13 1945 70
NM_001262316.5 NP_001249245.2 12 - 13 1964 70
NM_001306453.2 NP_001293382.1 12 - 13 1964 70
NM_001306454.3 NP_001293383.1 9 - 10 1444 70
NM_001306455.3 NP_001293384.1 9 - 10 1444 70

View Full Product Details