Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sulp-6
Entrez Gene ID
189085
Gene Name
protein-coding
Gene Aliases
CELE_W01B11.2
Chromosome Location
Chr.I: 3287369 - 3293171 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373791.2 NP_001360427.1 3 - 4 455 65
NM_058737.5 NP_491138.2 3 - 4 438 65

View Full Product Details