Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
inx-21
Entrez Gene ID
171930
Gene Name
Innexin
Gene Aliases
CELE_Y47G6A.1
Chromosome Location
Chr.I: 3611942 - 3618317 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058786.7 NP_491187.2 3 - 4 684 125

View Full Product Details