Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
inx-12
Entrez Gene ID
171944
Gene Name
Innexin-12
Gene Aliases
CELE_ZK770.3
Chromosome Location
Chr.I: 3753968 - 3759039 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058812.8 NP_491213.1 3 - 4 661 77

View Full Product Details